Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145152.4 | NP_660134.1 | 2 | 649 | ||
GenBank mRNA | AK043535.1 | - | 2 | 624 | ||
AY061858.1 | - | 2 | 622 | |||
BC058363.1 | - | 1 | 431 | |||
BC115652.2 | - | 1 | 457 | 40103158 |