Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013441.2 | NP_001013459.1 | 3 | 593 | ||
NM_183172.2 | NP_898995.1 | 3 | 593 | |||
GenBank mRNA | AK132369.1 | - | 3 | 497 | ||
AK137531.1 | - | 2 | 391 | |||
AY940666.1 | - | 3 | 562 | |||
AY940667.1 | - | 3 | 550 | |||
BC051080.1 | - | 3 | 590 | 6490136 |