Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smcr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085440.1 | NP_001078909.1 | 1 | 789 | ||
NM_175491.4 | NP_780700.1 | 1 | 789 | |||
GenBank mRNA | AK052754.1 | - | 1 | 777 | ||
AK083739.1 | - | 1 | 773 | |||
AK145010.1 | - | 1 | 775 | |||
AK164045.1 | - | 1 | 778 | |||
BC085095.1 | - | 1 | 419 | 30732606 |