Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4rl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177708.5 | NP_808376.1 | 2 | 1421 | ||
GenBank mRNA | AK033286.1 | - | 2 | 1419 | ||
AK140456.1 | - | 2 | 1422 | |||
AY250218.1 | - | 1 | 1132 | |||
BC030471.1 | - | 2 | 1625 | 5368560 |