Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tusc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177709.3 | NP_808377.1 | 3 | 1064 | ||
GenBank mRNA | AK048522.1 | - | 3 | 1064 | ||
AK158395.1 | - | 3 | 1027 | |||
BC119801.1 | - | 3 | 860 | 40107649 | ||
BC122875.1 | - | 3 | 860 | 40107655 |