Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030291.1 | NP_001025462.1 | 3 | 595 | ||
GenBank mRNA | AK144474.1 | - | 3 | 598 | ||
AK167678.1 | - | 3 | 598 | |||
BC141245.1 | - | 3 | 680 | 9055857 | ||
BC171967.1 | - | 3 | 680 | 9054253 |