Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001333.2 | NP_001001333.2 | 12 | 1685 | ||
NM_001146073.1 | NP_001139545.1 | 11 | 1490 | |||
GenBank mRNA | AK154237.1 | - | 12 | 1659 | ||
BC056487.1 | - | 8 | 3455 | |||
BC069960.1 | - | 11 | 1442 | 5007884 | ||
FM204885.1 | - | 11 | 1222 | |||
FM204886.1 | - | 11 | 1441 |