Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC068281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173416.3 | NP_775592.2 | 2 | 1991 | ||
GenBank mRNA | AK040407.1 | - | 2 | 1948 |