Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172152.2 | NP_742164.1 | 17 | 1784 | ||
GenBank mRNA | AK044239.1 | - | 16 | 3818 | ||
AK199037.1 | - | 1 | 44 | |||
AY156046.1 | - | 17 | 1784 | |||
BC137945.1 | - | 17 | 1755 | 8860964 | ||
BC171939.1 | - | 17 | 1755 | 9054225 |