Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033335.3 | NP_001028507.2 | 4 | 901 | ||
NM_001168294.1 | NP_001161766.1 | 4 | 942 | |||
NM_001168295.1 | NP_001161767.1 | 3 | 730 | |||
GenBank mRNA | AK138954.1 | - | 4 | 902 | ||
AK149697.1 | - | 4 | 944 | |||
AK150157.1 | - | 4 | 943 | |||
BC049975.1 | - | 3 | 720 | |||
BC139132.1 | - | 3 | 790 | 8862154 | ||
BC139133.1 | - | 3 | 790 | 8862155 |