Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021129.3 | NP_066952.1 | 10 | 933 | ||
GenBank mRNA | AB026723.1 | - | 10 | 856 | ||
AF092439.1 | - | 10 | 903 | |||
AF119665.1 | - | 10 | 903 | |||
AF154065.1 | - | 10 | 871 | |||
AF217186.1 | - | 10 | 826 | |||
AK058193.1 | - | 10 | 926 | |||
AK127102.1 | - | 3 | 2228 | |||
BC001022.2 | - | 10 | 908 | 3345496 | ||
BC061581.1 | - | 10 | 916 | 6526628 | ||
BC105034.1 | - | 10 | 899 | 8144037 | ||
BC105036.1 | - | 10 | 899 | 8144039 | ||
BC107882.1 | - | 10 | 867 | 6495107 | ||
EU794626.1 | - | 10 | 933 |