Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081128.3 | NP_001074597.1 | 13 | 1320 | ||
GenBank mRNA | AK037599.1 | - | 12 | 1255 | ||
AK142258.1 | - | 13 | 1303 | |||
BC145683.1 | - | 13 | 1207 | 40130986 | ||
BC145685.1 | - | 13 | 1207 | 40130988 |