Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004149.1 | NP_001004149.1 | 5 | 2213 | ||
GenBank mRNA | BC070399.1 | - | 5 | 2213 | 5694313 |