Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H3f3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008210.4 | NP_032236.1 | 4 | 831 | ||
GenBank mRNA | AK008405.1 | - | 3 | 732 | ||
AK037900.1 | - | 4 | 833 | |||
AK142749.1 | - | 1 | 55 | |||
AK150467.1 | - | 4 | 791 | |||
AK150468.1 | - | 4 | 791 | |||
AK150591.1 | - | 4 | 786 | |||
AK150928.1 | - | 4 | 786 | |||
AK151634.1 | - | 4 | 791 | |||
AK152293.1 | - | 4 | 786 | |||
AK153034.1 | - | 4 | 790 | |||
AK160677.1 | - | 4 | 790 | |||
AK162040.1 | - | 4 | 789 | |||
AK167879.1 | - | 4 | 790 | |||
AK168197.1 | - | 4 | 790 | |||
AK169608.1 | - | 4 | 786 | |||
BC002268.1 | - | 4 | 776 | 3495830 | ||
BC012687.1 | - | 4 | 780 | 4224280 | ||
BC083353.1 | - | 4 | 771 | 5714859 | ||
BC088835.1 | - | 4 | 768 | 6437687 | ||
BC106177.1 | - | 4 | 765 | 6511787 | ||
Z85979.1 | - | 4 | 779 |
Gene Symbol: Gm2099 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137674.1 | - | 3478 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC027275.1 | - | 2306 | Chr. - - [-] on Build |