Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146245.2 | NP_666357.2 | 4 | 1128 | ||
GenBank mRNA | AK044631.1 | - | 4 | 1129 | ||
BC032270.1 | - | 4 | 1134 | 5401567 |