Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033336.3 | NP_001028508.2 | 9 | 1351 | ||
NM_001163675.1 | NP_001157147.1 | 9 | 1333 | |||
NM_001163676.1 | NP_001157148.1 | 8 | 1126 | |||
GenBank mRNA | AK052778.1 | - | 9 | 1334 | ||
AK157723.1 | - | 9 | 1338 | |||
BC150822.1 | - | 9 | 1305 | 9087733 | ||
BC171974.1 | - | 8 | 1080 | 9054260 |