Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175502.3 | NP_780711.1 | 2 | 1371 | ||
GenBank mRNA | AK046420.1 | - | 2 | 1372 | ||
BC076589.1 | - | 1366 | ||||
BC117021.1 | - | 1 | 963 | 40126340 | ||
BC117025.1 | - | 1 | 963 | 40126344 |