Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153178.4 | NP_694818.3 | 8, 9 | 1199 | ||
GenBank mRNA | AB081472.1 | - | 8, 9 | 1061 | ||
AK220193.1 | - | 5, 6 | 564 | |||
BC024857.1 | - | 1, 2 | 44 | |||
BC064741.1 | - | 3, 4 | 316 | |||
BC096465.1 | - | 8, 9 | 1104 | |||
BC128379.1 | - | 8, 9 | 1052 | 40111876 | ||
BC129922.1 | - | 8, 9 | 1052 | 40111875 |