Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A4galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004150.3 | NP_001004150.1 | 3 | 691 | ||
NM_001170954.1 | NP_001164425.1 | 3 | 731 | |||
GenBank mRNA | AK139833.1 | - | 3 | 695 | ||
AK149186.1 | - | 3 | 647 | |||
AK162529.1 | - | 3 | 730 | |||
AY371179.1 | - | 3 | 650 | |||
BC138844.1 | - | 2 | 606 | 8861866 | ||
BC138845.1 | - | 2 | 606 | 8861867 | ||
DQ143885.1 | - | 1 | 425 |