Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172818.3 | NP_766406.2 | 12 | 2583 | ||
GenBank mRNA | AK030151.1 | - | 12 | 2576 | ||
AM690752.1 | - | 12 | 2240 | |||
BC116293.1 | - | 12 | 2224 | 40094952 | ||
BC116294.1 | - | 12 | 2219 | 40094955 |