Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd200r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207244.2 | NP_997127.1 | 3 | 429 | ||
GenBank mRNA | AK089262.1 | - | 5 | 724 | ||
AY230200.1 | - | 3 | 429 | |||
BC131946.1 | - | 3 | 419 | 40130223 | ||
BC131972.1 | - | 3 | 419 | 40130249 |