Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172825.3 | NP_766413.2 | 14 | 2084 | ||
GenBank mRNA | AK033593.1 | - | 14 | 2085 | ||
BC132302.1 | - | 14 | 1909 | 40130579 | ||
BC137963.1 | - | 14 | 1909 | 8860982 |
Set Membership: |
G-protein coupled receptor |