Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnldc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034866.1 | NP_001030038.1 | 4 | 324 | ||
GenBank mRNA | AK139523.1 | - | 4 | 210 | ||
BC158033.1 | - | 4 | 324 | 9007336 | ||
BC172002.1 | - | 4 | 324 |