Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033339.3 | NP_001028511.1 | 10 | 2187 | ||
GenBank mRNA | AK089273.1 | - | 10 | 1929 | ||
AK089533.1 | - | 10 | 1926 | |||
AK137491.1 | - | 10 | 1789 | |||
AK154458.1 | - | 10 | 2189 | |||
AK171493.1 | - | 10 | 1926 | |||
BC059059.1 | - | 8 | 2936 | |||
BC112379.1 | - | 10 | 1873 | 40059601 |