Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163013.1 | NP_001156485.1 | 14 | 2016 | ||
GenBank mRNA | BC037178.1 | - | 1 | 38 | ||
BC072654.1 | - | 9 | 1061 | |||
BC138263.1 | - | 14 | 2016 | 8861282 | ||
BC171951.1 | - | 14 | 2016 | 9054237 |