Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177983.2 | NP_817092.1 | 5 | 718 | ||
GenBank mRNA | AB209197.1 | - | 2 | 2371 | ||
AK127593.1 | - | 5 | 584 | |||
AK293141.1 | - | 6 | 512 | |||
AK312455.1 | - | 5 | 708 | |||
BC000057.2 | - | 5 | 567 | 3505271 | ||
BC022061.2 | - | 5 | 564 | 2988852 | ||
BT007454.1 | - | 5 | 457 | |||
CR457059.1 | - | 5 | 457 | |||
Y13936.1 | - | 5 | 481 |