Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam69c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173770.4 | NP_776131.1 | 4 | 991 | ||
GenBank mRNA | AK046504.1 | - | 4 | 992 | ||
BC119040.1 | - | 2 | 579 | 8733789 | ||
BC119042.1 | - | 2 | 579 | 8733791 |