Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42bpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033342.1 | NP_001028514.1 | 30 | 4020 | ||
GenBank mRNA | AK133191.1 | - | 7 | 1092 | ||
BC046418.1 | - | 13 | 1668 |