Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183179.1 | NP_899002.1 | 1 | 451 | ||
GenBank mRNA | AY238939.1 | - | 1 | 319 | ||
BC039042.1 | - | 1 | 451 | 5363843 | ||
BC046490.1 | - | 1 | 482 |