Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008709.1 | NP_001008709.1 | 3 | 413 | ||
NM_002708.3 | NP_002699.1 | 3 | 380 | |||
NM_206873.1 | NP_996756.1 | 2 | 248 | |||
GenBank mRNA | AK098311.1 | - | 5 | 1189 | ||
AK127616.1 | - | 4 | 597 | |||
AK313586.1 | - | 3 | 289 | |||
BC001888.1 | - | 3 | 301 | 3546588 | ||
BC004482.2 | - | 3 | 321 | 3689198 | ||
BC008010.1 | - | 3 | 321 | 3528742 | ||
BE278068.1 | - | 2 | 168 | |||
BE793459.1 | - | 3 | 383 | |||
BG480286.1 | - | 3 | 378 | |||
BM799599.1 | - | 2 | 187 | |||
BT006629.1 | - | 3 | 257 | |||
J04759.1 | - | 2 | 191 | |||
M63960.1 | - | 3 | 286 | |||
S57501.1 | - | 3 | 301 | |||
X70848.1 | - | 3 | 284 |