Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183046.1 | NP_898867.1 | 31 | 5229 | ||
GenBank mRNA | AK145779.1 | - | 17 | 3214 | ||
AY259532.1 | - | 23 | 4005 | |||
BC036147.1 | - | 13 | 2651 | |||
BC048954.1 | - | 12 | 1462 | |||
BC151061.1 | - | 31 | 5075 | 9087974 | ||
BC172148.1 | - | 31 | 5075 | 9054818 |