Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brinp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207583.2 | NP_997466.2 | 8 | 2795 | ||
GenBank mRNA | AK032297.1 | - | 8 | 2799 | ||
AK045801.1 | - | 8 | 2322 | |||
AK122549.1 | - | 8 | 2861 | |||
BC067038.1 | - | 8 | 3600 | |||
BC076572.1 | - | 8 | 2795 | 30363878 |