Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163314.2 | NP_001156786.1 | 20 | 1923 | ||
GenBank mRNA | AK138171.1 | - | 20 | 1863 | ||
CK392830.1 | - | 7 | 457 |