Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146250.2 | NP_666362.1 | 3 | 851 | ||
GenBank mRNA | BC032934.1 | - | 3 | 779 | 1363204 |
Set Membership: |
G-protein coupled receptor |