Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145838.1 | NP_665837.1 | 7 | 794 | ||
GenBank mRNA | AB059554.1 | - | 7 | 794 | ||
AK085105.1 | - | 8 | 931 | |||
AK156557.1 | - | 7 | 1331 | |||
AK172400.1 | - | 7 | 859 | |||
BC115912.1 | - | 7 | 749 | 40104779 | ||
BC118063.1 | - | 7 | 749 | 40104780 |