Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xirp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024618.2 | NP_001019789.1 | 7 | 5581 | ||
NM_001083919.1 | NP_001077388.1 | 7 | 5581 | |||
GenBank mRNA | DQ011666.1 | - | 7 | 5582 | ||
EF119716.1 | - | 7 | 5581 | |||
EF122140.1 | - | 6 | 5336 |