Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172856.3 | NP_766444.1 | 6 | 616 | ||
GenBank mRNA | AK028849.1 | - | 6 | 617 | ||
AK135798.1 | - | 6 | 718 | |||
AK151848.1 | - | 6 | 705 | |||
AK152417.1 | - | 6 | 686 | |||
BC057629.1 | - | 6 | 659 | 4224547 |