Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005342.2 | NP_001005342.1 | 5 | 620 | ||
GenBank mRNA | AB098743.2 | - | 5 | 620 | ||
AK043577.1 | - | 2 | 458 | |||
BC119263.1 | - | 4 | 494 | 8734012 |