Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290784.1 | NP_001277713.1 | 4 | 536 | ||
NM_178886.3 | NP_849217.2 | 4 | 539 | |||
GenBank mRNA | AK029683.1 | - | 4 | 437 | ||
AK032208.1 | - | 4 | 425 | |||
AK184960.1 | - | 2 | 132 | |||
AK198364.1 | - | 2 | 137 | |||
AK202757.1 | - | 2 | 148 | |||
BC138177.1 | - | 4 | 539 | 8861196 | ||
BC138178.1 | - | 5 | 537 | 8861197 | ||
BC145174.1 | - | 4 | 535 | 9053725 | ||
CN527209.1 | - | 4 | 456 |