Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290784.1 | NP_001277713.1 | 5 | 723 | ||
NM_178886.3 | NP_849217.2 | 5 | 726 | |||
GenBank mRNA | AK029683.1 | - | 5 | 624 | ||
AK032208.1 | - | 5 | 612 | |||
AK204565.1 | - | 1 | 74 | |||
AK204962.1 | - | 1 | 74 | |||
BC138177.1 | - | 5 | 726 | 8861196 | ||
BC138178.1 | - | 6 | 724 | 8861197 | ||
BC145174.1 | - | 5 | 722 | 9053725 | ||
CN527209.1 | - | 5 | 643 |