Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172857.2 | NP_766445.1 | 13 | 1907 | ||
GenBank mRNA | AK030132.1 | - | 13 | 1909 | ||
BC092241.1 | - | 11 | 1161 | 30670567 | ||
BC144750.1 | - | 12 | 1290 | 9053298 | ||
BC150662.1 | - | 13 | 1384 | 9087573 |