Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080944.3 | NP_001074413.2 | 19 | 2084 | ||
GenBank mRNA | AK086607.1 | - | 18 | 2073 | ||
AK155448.1 | - | 19 | 2094 | |||
AK173307.1 | - | 19 | 2084 | |||
BC130260.1 | - | 1 | 58 | |||
BC152337.1 | - | 1 | 58 |