Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080944.3 | NP_001074413.2 | 25, 26 | 3141 | ||
GenBank mRNA | AK155448.1 | - | 25, 26 | 3151 | ||
AK169967.1 | - | 25, 26 | 3169 | |||
BC130260.1 | - | 7, 8 | 1115 | |||
BC152337.1 | - | 7, 8 | 1115 |