Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzts3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291027.1 | NP_001277956.1 | 4 | 811 | ||
NM_001291028.1 | NP_001277957.1 | 4 | 469 | |||
NM_197945.4 | NP_922936.3 | 4 | 811 | |||
GenBank mRNA | AK148251.1 | - | 4 | 770 | ||
AK161711.1 | - | 4 | 765 | |||
BC049892.1 | - | 2 | 372 | |||
BC058280.1 | - | 3 | 393 | 6529102 | ||
DN175861.1 | - | 4 | 418 |