Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgapa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033348.3 | NP_001028520.2 | 4 | 393 | ||
GenBank mRNA | AK038838.1 | - | 4 | 301 | ||
AK157932.1 | - | 4 | 395 | |||
BC151040.1 | - | 4 | 365 | |||
BC151041.1 | - | 4 | 365 |