Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgapa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033348.3 | NP_001028520.2 | 6, 7 | 649 | ||
GenBank mRNA | AK038838.1 | - | 6, 7 | 557 | ||
AK157932.1 | - | 6, 7 | 651 | |||
BC151040.1 | - | 6, 7 | 621 | |||
BC151041.1 | - | 6, 7 | 621 |