Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014225.5 | NP_055040.2 | 3 | 514 | ||
XM_017026929.1 | XP_016882418.1 | 3 | 743 | |||
GenBank mRNA | AK090488.1 | - | 3 | 748 | ||
AK091387.1 | - | 3 | 294 | |||
AK290593.1 | - | 3 | 302 | |||
AK291932.1 | - | 3 | 297 | |||
AK293174.1 | - | 3 | 295 | |||
AK299012.1 | - | 3 | 271 | |||
AK307743.1 | - | 3 | 385 | |||
AK315700.1 | - | 3 | 330 | |||
AU128747.1 | - | 3 | 335 | |||
BC001537.2 | - | 3 | 260 | 2987938 | ||
BC026733.1 | - | 3 | 277 | |||
CR450340.1 | - | 3 | 219 | |||
DB246691.1 | - | 3 | 378 | |||
DC398468.1 | - | 3 | 516 | |||
J02902.1 | - | 3 | 363 |