Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178594.3 | NP_848709.2 | 4 | 569 | ||
GenBank mRNA | AK143276.1 | - | 4 | 570 | ||
AK145664.1 | - | 4 | 569 | |||
AY280973.1 | - | 4 | 469 | |||
AY322147.1 | - | 4 | 469 | |||
AY346099.1 | - | 4 | 549 | |||
BC032925.1 | - | 4 | 519 | 1477676 |