Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcna10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081140.1 | NP_001074609.1 | 2 | 467 | ||
GenBank mRNA | AK157965.1 | - | 2 | 468 | ||
BC137828.1 | - | 2 | 556 | 8860847 | ||
BC137829.1 | - | 2 | 556 | 8860848 | ||
BC158036.1 | - | 2 | 388 | 9007339 |