Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271899.1 | NP_001258828.1 | 10 | 1387 | ||
NM_001271900.1 | NP_001258829.1 | 11 | 1581 | |||
NM_001271948.1 | NP_001258877.1 | 10 | 1914 | |||
NM_181674.2 | NP_858060.2 | 10 | 1712 | |||
NM_181675.3 | NP_858061.2 | 10 | 1434 | |||
NM_181676.2 | NP_858062.1 | 9 | 1518 | |||
NM_181677.2 | NP_858063.1 | 11 | 1344 | |||
NM_181678.2 | NP_858064.1 | 11 | 1357 | |||
NR_073526.1 | 9 | 1829 | ||||
NR_073527.1 | 10 | 1466 | ||||
GenBank mRNA | AK056192.1 | - | 10 | 1394 | ||
AK289717.1 | - | 10 | 1363 | |||
AK294659.1 | - | 11 | 1490 | |||
AK295347.1 | - | 10 | 1358 | |||
AK314810.1 | - | 10 | 1711 | |||
BC031790.1 | - | 10 | 1602 | 4939981 | ||
BX647887.1 | - | 10 | 1448 | |||
M64930.1 | - | 9 | 1640 |